Heating mantles, multi-position, WHRE4 series, PILZ®
Leverancier: WINKLER
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
WHRE4E001000SS WHRE4E000500SS WHRE4E001000RC WHRE4E000100RC WHRE4E000100CH WHRE4E000100SS WHRE4E000250CH WHRE4E000250RC WHRE4E000250SS WHRE4E000500CH WHRE4E000500RC WHRE4E001000CH WHRE4E000100UK WHRE4E000250UK WHRE4E000500UK WHRE4E001000UK
WINKWHRE4E001000SSEA 1959 EUR
WINKWHRE4E001000SS WINKWHRE4E000500SS WINKWHRE4E001000RC WINKWHRE4E000100RC WINKWHRE4E000100CH WINKWHRE4E000100SS WINKWHRE4E000250CH WINKWHRE4E000250RC WINKWHRE4E000250SS WINKWHRE4E000500CH WINKWHRE4E000500RC WINKWHRE4E001000CH WINKWHRE4E000100UK WINKWHRE4E000250UK WINKWHRE4E000500UK WINKWHRE4E001000UK
Heating mantles, multi-position, WHRE4 series, PILZ®
Verwarmers Verwarmingsmantels
For round bottomed flasks. Max. 450 °C. Multiposition heating mantles with powder coated, chemically resistant metal housing and stainless steel front and cover plate. Mantles have crocheted outer jacket made from glass silk.
  • Integrated power selector per heating position, infinitely variable power 0 to 100%
  • Four heating zones
  • Detachable plug-in connection (cable length 0,5 m)
  • RCD personal safety plug (cable length 1,0 m)
Rod clamp STK1 per heating place (without rod).