Capillaries for infinityLab quick connect fittings
Leverancier: Agilent
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
5500-1172 5500-1173 5500-1170 5500-1171 5500-1174 5500-1175 5500-1176 5500-1177 5500-1178 5500-1179 5500-1180 5500-1181 5500-1182 5500-1183 5500-1230 5500-1231 5500-1258 5500-1259 5500-1260
558-0649EA 79 EUR
558-0649 558-0650 558-0647 558-0648 558-0651 558-0652 558-0653 558-0654 558-0655 558-0656 558-0657 558-0658 558-0659 558-0660 558-0672 558-0673 558-0680 558-0681 558-0682
Capillaries for infinityLab quick connect fittings
Slangen Chromatography Tubing
These capillaries are available in different lengths and internal diameters, including 0,075 mm ID, 0,12 mm ID, 0,17 mm ID and 0,25 mm ID to fit most UHPLC as well as HPLC applications.
  • Made of stainless steel
The 0,25 mm ID capillaries with a female connection are optimised for Waters Alliance systems.