Chemical resistant gloves, Camatex 451
Leverancier: KCL by Honeywell
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
451.10 451.7 451.8 451.9
KCHL451.10EA 317 EUR
KCHL451.10 KCHL451.7 KCHL451.8 KCHL451.9
Chemical resistant gloves, Camatex 451
Handschoenen
Camatex 451 gloves are made of comfortable, highly elastic natural latex with a cotton liner.
  • Excellent grip on wet surfaces
  • Remain comfortable in higher or lower temperatures, even when worn for extended periods
  • Fully coated - liquid proof
  • Excellent mechanical resistance
  • Roughened palm
Used in the food industry, construction industry, for harvesting, cleaning and steam blasting works, canteen kitchens, erection of scaffolding and in the beverage industry.
Certificaten: EN 388:2016 (2131X), EN ISO 374-1 Type A (KLMPST); Cat. III; AQL 1,5
Food contact approved.