Nitrile gloves, SemperCare®
Leverancier: SEMPERIT TECHN PROD
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
826781633 826781635 826781637 826781639
SEMP826781633EA 26.4 EUR
SEMP826781633 SEMP826781635 SEMP826781637 SEMP826781639
Nitrile gloves, SemperCare®
Handschoenen
The ambidextrous disposable SemperCare® Nitrile combines superb barrier protection, comfort, fit, and feel while providing the tensile strength necessary for demanding environments.
  • Easy donning, improved fit and comfort
  • Strong and more durable
  • Powder- and latex-free
  • Beaded cuff
  • Textured fingers and palm for safe grip, even in wet environments
  • Smart Dispense Technology, Single Glove Delivery® for fewer wrinkles
Appropriate for handling cytotoxic chemicals. Safe for use with sensitive electronics.