Latexometers
Leverancier: Amarell
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
H860580 H860582
AMARH860580EA 0 EUR
AMARH860580 AMARH860582
Latexometers
Hydrometers
These instruments are calibrated in grams of dry rubber per litre of latex.
  • Without thermometer
  • Paper scale with well visible print
  • Shot weighted body
Accuracy: ±1%
Reference temperature: 28,9 °C