BrightVision+ Anti-IgG (Poly-AP)

Leverancier: IMMUNOLOGIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

DPVO110AP DPVO500AP DPVO999AP
DPVO110APEA 1330 EUR
DPVO110AP DPVO500AP DPVO999AP
BrightVision+ Anti-IgG (Poly-AP)
Antilichamen
BrightVision histostaining reagents utilise a novel controlled polymerisation technology to prepare polymeric AP-linker antibody conjugates. Compared to conventional biotin-streptavidin based detection kits, BrightVision histostaining kits have the advantages of higher amplification power, biotin free and more consistent immunostaining outcomes on archival tissues and on difficult-to-work antibodies. These advantages would bring to laboratories the benefit of more accurate result, faster turn-around, less trouble shooting and better costs-saving.

  • High-quality secondary antibody for IHC applications, AP labelled

Kit contains Poly-AP-anti-Rabbit/Mouse IgG ready to use.

Biotin-free

Certificaten: Meets CE/IVD standards.

Bestelinformatie: Packages above 110 ml contain small empty bottles so users can create smaller working volumes and prevent possible contamination of the whole lot.

Leveringsinformatie: Shipped with blue ice. Store between 2 to 8 °C.

Opgelet: Irritating to eyes, respiratory systems and skin.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR