Multi-element standard solutions for ion chromatography
Leverancier: ACTIVE MOTIF
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1521.1K.W.L1 1521.1K.W.L25 1521.K1.W.L1 1521.K1.W.L25
ACSD1521.1K.W.L1EA 182 EUR
ACSD1521.1K.W.L1 ACSD1521.1K.W.L25 ACSD1521.K1.W.L1 ACSD1521.K1.W.L25
Multi-element standard solutions for ion chromatography
Standaarden Anionische standaarden Anionische standaarden met meerdere elementen
This standard solution is intended for ion chromatography.
Multi element standards for Br⁻, Cl-, F⁻, NO₂⁻, NO₃⁻, PO₄³⁻, SO₄²⁻ diluted in H₂O.