Centrifuge bottles, PPCO/PC, Fiberlite™, Sorvall™
Leverancier: SORVALL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
010-1493 010-1494 010-1492 010-1495 010-1491
SORV010-1493EA 333 EUR
SORV010-1493 SORV010-1494 521-2669 SORV010-1495 521-2668
Centrifuge bottles, PPCO/PC, Fiberlite™, Sorvall™
Flessen Centrifugeerflessen
These Fiberlite™ bottle systems combine a Nalgene bottle body with a newly designed Fiberlite cap closure.
For use with Thermo Scientific™ Sorvall™ LYNX Superspeed and Bios A large-capacity centrifuges.