Bottles, narrow neck, round, with thread, 321
Leverancier: Kautex
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
321-86710 321-86711 321-86712
215-3245EA 555 EUR
215-3245 215-3246 215-3247
Bottles, narrow neck, round, with thread, 321
Flessen Kunststof flessen
HDPE. With PA barrier layer. White.
  • Special packaging
  • Barrier layer
  • Leakproof
  • Not suitable for food
  • Halal certified
High-quality packaging with barrier layer of polyamide for highly volatile or intense smelling media with high permeation rates.