Thread adapter, GL PTFE , BOLA
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
D537-04 D537-08 D537-16 D537-12
BOHLD537-04EA 41.8 EUR
BOHLD537-04 BOHLD537-08 BOHLD537-16 BOHLD537-12
Thread adapter, GL PTFE , BOLA
Fittingen en connectoren
Straight transition thread adapter made of PTFE with two GL-threaded connections.
  • Chemical resistant
  • Vacuum suitable
  • Temperature resistant from –200 to +250 °C