GC septa, thermolite® PLUS
Leverancier: Restek
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
23868 23864 23877
REST23868EA 107 EUR
REST23868 REST23864 REST23877
GC septa, thermolite® PLUS
Tussenschoten
These septa are preconditioned, GC-FID tested and ready to use.
  • Usable to 350 °C inlet temperature
  • Ultra-low bleed minimises background signal
  • Plasma coating eliminates sticking
  • Precision molded for consistent, accurate fit