Pocket thermometers, with clip
Leverancier: Amarell
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
G15602-FL G15606-FL
AMARG15602-FLEA 44.1 EUR
AMARG15602-FL AMARG15606-FL
Pocket thermometers, with clip
Thermometers Analoge thermometers
The thermometers with Hg filling have a blue reflecting capillary.
  • Pen size
  • Total immersion
  • Enclosed scale
Leveringsinformatie: All pocket thermometers supplied in nickel plated double receptacle.
Order Now
Specificaties
Environmentally Preferable