Microsampling tools
Leverancier: PerkinElmer
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
N1870151 N9302603 N9302605 N9302609 N9302610 N9302622
PERCN1870151EA 738 EUR
PERCN1870151 PERCN9302603 PERCN9302605 PERCN9302609 PERCN9302610 PERCN9302622
Microsampling tools
Gereedschapset
This microsampling tool kit is suitable for FTIR microscopy.
Leveringsinformatie: Microsampling tool kit includes dissecting forceps, roller knife, straight micro probe, aluminum probe handle, pin vise.