Vial racks
Leverancier: MACHEREY-NAGEL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
702502 702503 702504
MANA702502EA 17.1 EUR
MANA702502 MANA702503 MANA702504
Vial racks
Rekken Rekken voor chromatografievials
These chromatography vial racks are used for a safe standing position of the vials on the lab bench.
  • For 1,5 and 4 ml vials or for 10 and 20 ml headspace vials
  • Made from blue, chemically resistant PP
  • Empty vial racks are stackable