Protease and phosphatase inhibitor, Protease-PhosphataseArrest™
Leverancier: G Bioscienes
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
786-889 786-872 786-871 786-870
GENO786-889EA 889 EUR
GENO786-889 GENO786-872 GENO786-871 GENO786-870
Protease and phosphatase inhibitor, Protease-PhosphataseArrest™
Celcultuursupplementen en -additieven
Protease-PhosphataseArrest™ provides full protection of protein samples from proteases and phosphatases released during the preparation of cell and tissue lysates. Protease-PhosphataseArrest™ contains both irreversible and reversible protease inhibitors to inhibit serine, cysteine and other proteases along with five phosphatase inhibitors that target all the phosphatase categories: serine/threonine (Ser/Thr) specific, tyrosine specific and dual specificity phosphatases.
  • Complete protection of proteins – contains both protease and phosphatase inhibitors
  • Optional metalloprotease Inhibitor – 0,5M EDTA is supplied separately
  • Supplied as two vials of each cocktail in a ready to use format at 100X concentration
Application: Protects protein samples from protease and phosphatase activites during extraction and purification from cells and tissues.
Bestelinformatie: Supplied as a vial of ProteaseARREST™ and a vial of PhosphataseARREST™.