Microscope stands, Dino-Lite, RK-06A / RK-06-AE
Leverancier: Dino-Lite / IDCP B.V.
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
RK-06A RK-06-PL RK-06AE RK-06F
IDCPRK-06AEA 243 EUR
IDCPRK-06A IDCPRK-06-PL IDCPRK-06AE IDCPRK-06F
Microscope stands, Dino-Lite, RK-06A / RK-06-AE
Staafstatieven
These mid-range tabletop stands with square metal base are either designed for general purposes or for ESD applications. They can be used with all handheld Dino-Lite microscopes.
  • Constructed of stainless steel and lightweight aluminium
  • Adjustable horizontal arm has a 15 cm range with 360-degree rotation
  • Smooth focus adjustment and quick vertical movement: Ease the task of focusing when using Dino-Lite microscopes
  • Ease-of-assembly and adaptable extension arm design adds further mobility and flexibility to user’s applications
The RK-06-AE comes with an ESD (electrostatic discharge) safe base plate instead of the standard RK-06A base plate.
For users that require additional working distance, the optional RK-06-PL extension pole is available. It is required when working with the Dino-Lite Long Working Distance and Extra Long Working Distance microscopes.
Leveringsinformatie: Supplied with additional horizontal arm RK-10-EX.