Bottles, stainless steel
Leverancier: Sampling Systems
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
A836-1000 A836-500 A836-250
SAMPA836-1000EA 390 EUR
SAMPA836-1000 SAMPA836-500 SAMPA836-250
Bottles, stainless steel
Flessen Metalen flessen
Stainless steel bottles are ideal for storing and transporting powders and liquids. The high-quality, crevice-free finish makes these containers ideal for the most demanding applications.
  • 316L stainless steel body and lid
  • Multi-purpose containers
  • Robust construction
  • All welds are ground and polished smooth so its ideal for hygienic industries
  • Fitted with a 2" tri-ferrule leakproof lid system
The containers come as standard with a removable, easy clean silicone seal.
Certificaten: Seal is food grade and meets all the latest FDA and EC compliance standards.
Leveringsinformatie: Supplied with lid, silicone gasket (FDA compliant), and clampband.