Centrifuge bottles for Sorvall™ BIOS16
Leverancier: SORVALL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
75003870 75003871
521-2674EA 566 EUR
521-2674 521-3421
Centrifuge bottles for Sorvall™ BIOS16
Flessen Centrifugeerflessen
Process a 16 L harvest in a single run with the Thermo Scientific™ 2000 ml bottle, the ideal solution for the separation of large volume harvests. Designed for use in Thermo Scientific™ Sorvall™ BIOS 16 Bioprocessing Centrifuge and 6 x 2000 ml or 8 x 2000 ml Swing Bucket Rotor.