4-(Fmoc-hydrazino)benzoylaminomethyl resin, beads, NovaGel™, Novabiochem®
Leverancier: Novabiochem (Part of Merck)
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
8.55037.0001 8.55037.0025 8.55037.0005
01-64-0377-1EA 97 EUR
01-64-0377-1 01-64-0377-25 01-64-0377-5
4-(Fmoc-hydrazino)benzoylaminomethyl resin, beads, NovaGel™, Novabiochem®
4-(Fmoc-hydrazino)benzoylaminomethyl resin
The PEG-PS resin is prepared from aminomethyl (styrene -1% DVB). 100 - 200 mesh.
Order Now
Specification Test Results
Color (Visual) Yellowish to Beige
Appearance of Substance (Visual) Beads
Loading (Photometric determination of the Fmoc-chromophore liberated upon treatment with DBU/DMF) 0.40 - 0.80 mmol/g