Resin seeker filter plates, 96-well
Leverancier: TOSOH Bioscience
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
0045509 0045501 0045502 0045503 0045506 0045507 0045510 0045511 0045520
TOYO0045509EA 635 EUR
TOYO0045509 TOYO0045501 TOYO0045502 TOYO0045503 TOYO0045506 TOYO0045507 TOYO0045510 TOYO0045511 TOYO0045520
Resin seeker filter plates, 96-well
Microplaten Filtreerplaten
Resin seeker 96-well plates are disposable filter plates packed with TOYOPEARL resins available in several configurations for antibody affinity, ion exchange, HIC, and mixed-mode chromatography.
  • Can be operated manually using a multi-channel pipette or in an automated system designed for high throughput screening
Resin seeker 96-well plates can be used to screen multiple steps of the purification process including binding, wash and elution conditions in addition to resin selectivity, binding kinetics, purity and recovery of target molecule.