Safety googles, 505UP
Leverancier: UNIVET
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
505U.00.00.00 505U.00.00.11
UNVT505U.00.00.00EA 85.4 EUR
UNVT505U.00.00.00 UNVT505U.00.00.11
Safety googles, 505UP
Ruimzichtbrillen
Extremely light and encircling goggles.
  • Elastic structure, extremely flexible and resistant
  • New temple for a better ergonomy
  • Optimal comfort and fit thanks to the soft adjustable nosepad
Certificaten: EN 166
Leveringsinformatie: Neck cord included in the packaging.