Safety goggles, 506UP
Leverancier: UNIVET
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
506U.03.00.00 506U.03.02.00
UNVT506U.03.00.00EA 113 EUR
UNVT506U.03.00.00 UNVT506U.03.02.00
Safety goggles, 506UP
Ruimzichtbrillen
506UP presents technical and aesthetic improvements. Lens available with VANGUARD PLUS coating.
  • Panoramic wrapping lens available with AF+ and AS+ coatings
  • Temples regulation in length and inclination
  • Soft terminal tips to eliminate local pressure
  • Optimal comfort and fit thanks to the soft adjustable nosepad
  • New range of colours
Weight: 29 g