Dropping funnels, cylindrical, with double jacket
Leverancier: NEUBERT VOLUME GLASSWAERE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1596-20-0119 1596-20-0050
NEUB1596-20-0119EA 679 EUR
NEUB1596-20-0119 NEUB1596-20-0050
Dropping funnels, cylindrical, with double jacket
Trechters Druppeltrechters
Borosilicate glass 3.3, with PTFE tap.
  • Graduated
  • With drop counter
  • with GL 14 olives (PE)
  • With pressure compensation