LED head torch
Leverancier: Portwest
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PRTWPA50SIREA 8 EUR
PRTWPA50SIR
LED head torch
Lampen Zaklampen
Head light with tilt control, made of ABS, used to aim the light where it is needed most. Designed for a wide array of tasks, from security to everyday use.
  • Ideal for keeping hands free for work
  • Eight super-bright LEDs - 40 Lumens of brightness and 10 hours of run time
  • Tilt control for different working positions
Beam distance: 15 m (high), 10 m (medium).
Certificaten: RoHS certified.
Leveringsinformatie: Three AAA batteries included.