Test sieves, 305×40 mm
Leverancier: Retsch GmbH
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
60.259.000025 60.259.000032 60.259.000038 60.259.000045 60.259.000053 60.259.000063 60.259.000075 60.259.000090 60.259.000106 60.259.000125 60.259.000150 60.259.000180 60.259.000212 60.259.000250 60.259.000300 60.259.000355 60.259.000425 60.259.000500 60.259.000600 60.259.000710 60.259.000850 60.259.001000 60.259.001180 60.259.001400 60.259.001700 60.259.002000 60.259.002360 60.259.002800 60.259.003350 60.259.004000 60.259.004750 60.259.005600 60.259.006300 60.259.006700 60.259.008000 60.259.009500 60.259.011200 60.259.012500 60.259.013200 60.259.016000 60.259.019000 60.259.022400 60.259.025000 60.259.026500 60.259.031500 60.259.037500 60.259.045000 60.259.050000 60.259.053000 60.259.063000 60.259.075000 60.259.090000 60.259.100000 60.259.106000 60.259.125000
510-6201EA 835 EUR
510-6201 510-6202 510-6203 510-6204 510-6205 510-6206 510-6207 510-6208 510-6209 510-6210 510-6211 510-6212 510-6213 510-6214 510-6215 510-6216 510-6217 510-6218 510-6219 510-6220 510-6221 510-6222 510-6223 510-6224 510-6225 510-6226 510-6227 510-6228 510-6229 510-6230 510-6231 510-6232 510-6233 510-6234 510-6235 510-6236 510-6237 510-6238 510-6239 510-6240 510-6241 510-6242 510-6243 510-6244 510-6245 510-6246 510-6247 510-6248 510-6249 510-6250 510-6251 510-6252 510-6253 510-6254 510-6255
Test sieves, 305×40 mm
Zeven Testzeven
Sieves made from 304 stainless steel are individually laser engraved with clear and precise labelling for full traceability.
  • Sieve bodies are made in one piece and the absence of hollow spaces prevents cross-contamination
  • High corrosion resistance and easy to clean
  • Maximum stability and optimal sealing of the sieve stack, 15% less weight than traditional sieves
Sieve meshes, frames and labeling comply with standards. The sieves are tested five times and are delivered with test report.
Certificaten: An individual inspection certificate for test materials monitoring according to ASTM E 11 is available
Bestelinformatie: Sieves and accessories of other dimensions are available on request. Also available with perforated plate, round or square.