Safety shoes, slip-on, Uni6, 1740
Leverancier: Abeba
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1740/47 1740/41 1740/48 1740/42 1740/43 1740/44 1740/45 1740/46 1740/35 1740/36 1740/37 1740/38 1740/39 1740/40
114-5970EA 96.3 EUR
114-5970 114-5964 114-5971 114-5965 114-5966 114-5967 114-5968 114-5969 114-5958 114-5959 114-5960 114-5961 114-5962 114-5963
Safety shoes, slip-on, Uni6, 1740
Schoenen en laarzen
Anti-static safety shoes made from microfibre upper and a fuel resistant TPU outsole with high slip resistance, animal fats and oil resistance.
  • Breathable lining with silver ions
  • Replaceable soft comfort insole
  • Instep area with elastic band
  • Foamed PU midsole
  • Sanitised treatment reduces odour and bacterial growth
  • Protective steel toecap
  • Washable at 30 °C
Suitable for use in kitchens.
Colour: White
Certificaten: DGUV 112-191; HACCP; EN ISO 20345:2011; S2, SRC