Blue filter for myECL™ imager
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Pierce
PIER62241EA 1330 EUR
PIER62241
Blue filter for myECL™ imager
Fotodocumentatiesystemen
Thermo Scientific™ 445 to 480 nm bandpass filter is a threaded, 50 mm diameter glass filter. This product is used for the addition of a blue filter to one of two open positions in the myECL Imager for detection of blue-fluorescent dyes and stains.