Multiple assay microfluidic chips
Leverancier: PerkinElmer
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PERK760624EA 0 EUR
PERK760624
Multiple assay microfluidic chips
Diagnostische systemen Clinical Instruments
Microfluidic chips are designed to be used with multiple assays and can be reused for additional cost benefit.