Low protein binding collection tubes, Pierce™
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
88380 88379
PIER88380EA 248 EUR
PIER88380 PIER88379
Low protein binding collection tubes, Pierce™
Buisjes Dialysebuisjes
Pierce™ low protein binding microcentrifuge tubes help to minimise protein sample loss during sample manipulation and are ideal in applications where processing steps are extensive.
  • Durable, can able to tolerate centrifuge speeds up to 20000 RCF
  • Compatible, suitable for IP and mass spectrometry applications
  • RNase/DNase-free
These tubes are conveniently packed to avoid contamination.