Safety shoes, slip-on, cream
Leverancier: Jallatte
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
00ABI24/35 00ABI24/36 00ABI24/37 00ABI24/38 00ABI24/39 00ABI24/40 00ABI24/41 00ABI24/42 00ABI24/43 00ABI24/44 00ABI24/45 00ABI24/46 00ABI24/47 00ABI24/48
JALC00ABI24/35EA 80.4 EUR
JALC00ABI24/35 JALC00ABI24/36 JALC00ABI24/37 JALC00ABI24/38 JALC00ABI24/39 JALC00ABI24/40 JALC00ABI24/41 JALC00ABI24/42 JALC00ABI24/43 JALC00ABI24/44 JALC00ABI24/45 JALC00ABI24/46 JALC00ABI24/47 JALC00ABI24/48
Safety shoes, slip-on, cream
Schoenen en laarzen
Anti-static shoes made from water resistant microfiber upper. Washable in soap and water.
  • Dual density PU sole , outsole in PU white and shock-absorbing system
  • Resistant to oils and hydrocarbons
  • Footbed single-block structure made of soft polyurethane with absorbent
  • Breathable fabric textile, anatomical and antistatic
  • Vamp lined in non-woven fabric
  • Protective aluminium toecap (200 J)
Padded collar with compact flexible foam for extra comfort and better ankle and achilles tendon protection. Snug fit has elastic band underneath protective tongue. Upper lined in technical textiles OVER DRY and resistant.
Colour: Cream
Certificaten: DGUV 112-191; EN ISO 20345 : 2011; S2, SRC