Heat Transfer Liquids, Kryo 30
Leverancier: Heidolph
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HEID569-00610-00EA 559 EUR
HEID569-00610-00 HEID569-00611-00
Heat Transfer Liquids, Kryo 30
Vloeistoffen voor circulatiebaden
Non-flammable thermal fluid for heating and low-temperature range. Thermal fluid for Hei-CHILL chillers.
  • Completely water soluble
  • Monoethylene glycol/water mixture containing inhibitors
  • Density: 1,082 kg/m³
Recommended temperature range: –30 to +90 °C.