DNA specimen collection cards, Ahlstrom-Munksjö GenReleaz™
Leverancier: Ahlstrom-Munksjö
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
8.295.0001.B-N 8.295.0000.B-N 8.295.0002.B-N 8.296.0001.B-N 8.296.0000.B-N 8.296.0002.B-N
730-0623EA 640 EUR
730-0623 730-0530 730-0625 730-0654 730-0627 730-0656
DNA specimen collection cards, Ahlstrom-Munksjö GenReleaz™
Opslagsystemen voor nucleïnezuren
GenReleaz™ and GenReleaz™ Colour specimen collection cards are specifically developed for collection, transport, preservation and fast and efficient elution of DNA from biological fluids stored at ambient temperature.
  • Easy sample collection
  • Stabilisation of DNA inside fiber-based material
  • Long-term ambient preservation of DNA
  • Fast and simple DNA elution
  • Less than 30 minutes processing time
  • Cost saving with no need of cold-chains
One of the key features of GenReleaz™ is the method for recovering DNA: efficient elution process eliminates the need to use DNA extraction kits.
Can be used in multiplex PCR and quantitative PCR analysis, STR and SNP analysis, biobanking, pharmacogenomics, infectious diseases screening, genotyping, genetic identification, transgenic detection.
Certificaten: ISO 18385:2016