Microtitre tubes, 1,2 ml
Leverancier: BIOZYM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
710410 710413
BZYM710410EA 0 EUR
BZYM710410 BZYM710413
Microtitre tubes, 1,2 ml
Buisjes Microbuisjes
These standard 1,2 ml microtiter tubes are made of polypropylene and have a labeling field.
  • Autoclavable
  • Freezable
  • Can be closed with PlugStrip cap strips
The standard racks can be refilled easily and inexpensively with the reload system. Rack version works on Beckman, Tecan and Tomtec systems.
For individual vessels, the PlugStrip closures can be individually tailored. All microtiter tubes have an additional filling space that allows sample dilutions and/or sample mixing prior to transfer to microplates.
Possible applications include: HTLV-IIIMore Product Information