Cold Resistant Gloves, Cryo Gloves
Leverancier: Thermo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
189562 189561
HERA189562EA 291 EUR
HERA189562 HERA189561
Cold Resistant Gloves, Cryo Gloves
Handschoenen
Safely handle samples from ultra-low and cryogenic storage with these cryo gloves which offer a high level of thermal protection.
  • Comfortable, lightweight, breathable protection
  • Side release buckles for quick on/off
Bestelinformatie: Available in a variety of sizes.