Safety shoes, lace-up, Uni6, 31629
Leverancier: Abeba
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
31629/42 31629/43 31629/47 31629/46 31629/39 31629/40 31629/41 31629/35 31629/36 31629/37 31629/38 31629/48 31629/44 31629/45
ABEB31629/42EA 107 EUR
ABEB31629/42 ABEB31629/43 ABEB31629/47 ABEB31629/46 ABEB31629/39 ABEB31629/40 ABEB31629/41 ABEB31629/35 ABEB31629/36 ABEB31629/37 ABEB31629/38 ABEB31629/48 ABEB31629/44 ABEB31629/45
Safety shoes, lace-up, Uni6, 31629
Schoenen en laarzen
Safety shoes are made of breathable textile with pattern.
  • Water-repellent
  • Lining with silver ions
  • Metal-free penetration safety
  • Suitable for kitchen
  • ORTHOsat sole possible
  • Anti-slip TPU outsole
  • Sanitized treated
These shoes has soft PU midsole and interchangeable soft comfort insole.
Certificaten: EN ISO 20345: 2011 S3, DGUV 112-191, SRC.