Chemical resistant gloves, uvex u-fit strong N2000
Leverancier: UVEX
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
6096207 6096208 6096209 6096210 6096211
UVEX6096207EA 20 EUR
UVEX6096207 UVEX6096208 UVEX6096209 UVEX6096210 UVEX6096211
Chemical resistant gloves, uvex u-fit strong N2000
Handschoenen
These reinforced disposable gloves are made from 0,20 mm nitrile rubber and provide impressive mechanical strength. They are flexible and robust and provide good protection against many chemicals and splashes.
  • Very high mechanical strength
  • Outstanding tactile feel
  • Good grip
  • Silicone-free according to imprint test
Typical application include: Laboratories, chemical industry, precision assembly work, painting work, cleaning and food industry.
Certificaten: EN ISO 374-1:2016 Type A (JKLOPST)
Food contact approved