Flask clamps for shakers
Leverancier: OHAUS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
30400090 30400091 30400092 30400093 30400094 30400095 30400096 30400097 30400098 30400099 30400100 30400101 30400102 30400103
OHAU30400090EA 53.9 EUR
OHAU30400090 OHAU30400091 OHAU30400092 OHAU30400093 OHAU30400094 OHAU30400095 OHAU30400096 OHAU30400097 OHAU30400098 OHAU30400099 OHAU30400100 OHAU30400101 OHAU30400102 OHAU30400103
Flask clamps for shakers
Klemmen Klemmen voor algemeen gebruik
Stainless steel or PVC. PVC clamps are designed for use with polycarbonate flasks, and are autoclavable. Stainless steel are used for glass or polycarbonate flask materials.
Leveringsinformatie: Includes the hardware required to mount to shakers.