Gel filtration media, Sephadex® G-50
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sephadex™
SIALG50150-10GEA 124 EUR
SIALG50150-10G SIALG5050-10G SIALG5080-50G SUPLG50150-50G SIALG5050-50G SIALG5080-10G SIALG5050-100G
Gel filtration media, Sephadex® G-50
Eiwitzuivering Protein Purification Resins
Sephadex® G-50 is a gel filtration medium used in affinity chromatography, protein chromatrography and gel filtration chromatography.
  • Swelling from 1 g to (9 to 11 ml)
It has been used to investigate effects from snake venom, isolate lectins from various species and to demonstrate the positive influence of 20% ethanol on proteinase desorption.
Verpakking: Packed in poly bottle.