Wringers for mops
Leverancier: CONTEC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
2710 2756 2690 2689 2691 2694
141-0959EA 1040 EUR
141-0959 141-1434 141-1433 141-1432 CONT2691 CONT2694
Wringers for mops
Moppen
Variety of wringers for curved and flat mops.
  • Made of stainless steel
  • Can be used with curved, flat, and string mops
  • Ideal for use with Contec’s 15, 20, and 25 L bucket systems
  • Fully autoclavable