Spill control mats for instrument storage, Aesculap®
Leverancier: Aesculap
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
JF938 JF939 JF940 JF941
AESCJF938EA 45.7 EUR
AESCJF938 AESCJF939 AESCJF940 AESCJF941
Spill control mats for instrument storage, Aesculap®
Absorptiematerialen en bescherming tegen morsen
These mats are made of silicone mesh and ideal for spill free storage of instruments.
These mats possess good drying characteristics and gentle storage during mechanical processing.
Opgelet: Not suitable for hot air sterilisation.