Cut resistant gloves, uvex unidur 6648
Leverancier: UVEX
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
6093206 6093207 6093208 6093209 6093210 6093211
UVEX6093206EA 85 EUR
UVEX6093206 UVEX6093207 UVEX6093208 UVEX6093209 UVEX6093210 UVEX6093211
Cut resistant gloves, uvex unidur 6648
Handschoenen
These HPPE/elastane cut protection gloves with PU coating on palm and fingertips provide a cut protection level of 3/B. They are extremely sensitive and exceptionally well suited to fine assembly work in which a high level of tactility and cut protection are required.
  • Outstanding mechanical abrasion resistance
  • Good grip in dry and slightly damp areas
  • Good tactile feel, high flexibility and good wearer comfort
Areas of application: Automotive industry, repair work, light to medium metal processing and packaging.
Certificaten: EN 388:2016 (4342B).