Industrial gloves, uvex unipur 6639
Leverancier: UVEX
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
6024810 6024811 6024809 6024808 6024807
UVEX6024810EA 21.7 EUR
UVEX6024810 UVEX6024811 UVEX6024809 UVEX6024808 UVEX6024807
Industrial gloves, uvex unipur 6639
Handschoenen
These lightweight, sensitive and dirt-resistant polyamid safety gloves with PU coating are ideal for precision mechanical work.
  • Good mechanical abrasion resistance
  • Good grip in dry and slightly damp areas
  • Outstanding tactile feel
  • Very good fit
Typical applications include: Fine assembly work, precision work, inspection and sorting.
Certificaten: EN 388:2003 (4131)