Industrial gloves, uvex unipur 6631
Leverancier: UVEX
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
6094406 6094407 6094408 6094409 6094410 6094411
UVEX6094406EA 26 EUR
UVEX6094406 UVEX6094407 UVEX6094408 UVEX6094409 UVEX6094410 UVEX6094411
Industrial gloves, uvex unipur 6631
Handschoenen
These lightweight PA safety gloves with PU coating on palm and fingertips are suitable for mechanical precision work in dry or slightly damp working environments.
  • Good mechanical robustness and abrasion resistance
  • Outstanding tactile feel
  • Highly flexibile
  • Very good fit
Ideal for precision assembly work, precision engineering, inspection, sorting.
Certificaten: EN 388:2016 (4141X)