Industrial gloves, uvex phynomic wet plus
Leverancier: UVEX
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
6006111 6006110 6006109 6006108 6006107
UVEX6006111EA 59.2 EUR
UVEX6006111 UVEX6006110 UVEX6006109 UVEX6006108 UVEX6006107
Industrial gloves, uvex phynomic wet plus
Handschoenen
These polyamide/elastane gloves with aqua-polymer foam coating on palm, fingertips and parts of the back of the hand offers outstanding protection against humidity. They fit like a second skin and are particularly light and flexible, without making the hands sweat.
  • Water-repellent coating and liner
  • Outstanding mechanical abrasion resistance
  • High level of breathability
  • Excellent skin compatibility
  • Very good tactile feel when assembling parts
  • Very good grip in damp, wet and oily areas
Ideal for use in more demanding outdoor areas of application. Typical applications include: Fine assembly work or assembly work, general maintenance work and precision work.
Certificaten: EN 388:2016 (3131X)
OEKO-Tex® Standard 100