Industrial gloves, uvex profi ergo XG20
Leverancier: UVEX
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
6020807 6020808 6020809 6020810
UVEX6020807EA 75.2 EUR
UVEX6020807 UVEX6020808 UVEX6020809 UVEX6020810
Industrial gloves, uvex profi ergo XG20
Handschoenen
These cotton interlock gloves feature a special NBR and Xtra Grip coating on the palm and back of the hand. Ideal for use in oily and greasy areas of application.
  • Excellent mechanical abrasion resistance thanks to the multi-layer design for increased service life
  • Outstanding grip in damp, wet and oily areas
  • Excellent wearer comfort due to the high water vapour absorption of the cotton lining
  • Good tactile feel
  • Ergonomic fit, highly flexible
Typical applications include: Assembly, inspection, maintenance, light to medium metal processing, concrete/construction work and outdoor activities.
Certificaten: EN 388:2016 (3121X)
OEKO-Tex® Standard 100