Bump caps, uvex u-cap sport vent
Leverancier: UVEX
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
9794.421 9794.420 9794.430
UVEX9794.421EA 28 EUR
UVEX9794.421 UVEX9794.420 UVEX9794.430
Bump caps, uvex u-cap sport vent
Hoofdbescherming Stootpetten
These caps with airy mesh fabric feature an innovative hard shell element for reliable impact protection.
  • Protect against abrasions and impact injuries
  • Airy mesh fabric for improved wearer comfort, especially in warm environments
  • Hard shell element in 'armadillo' design with honeycomb structure and shock-absorbing elements
  • Integrated reflective piping on the brim and textile components
  • Can be quickly and easily fitted to the head using the adjustable Velcro® fastening
  • With short peak for a wider field of upward vision
The armadillo design provides additional flexibility and ergonomically adapts to different head shapes and sizes. The extended cut-out near the ear makes it easier to wear earmuffs.
Certificaten: EN 812