Lugol's iodine solution stabilised for Gram staining, Sigma-Aldrich®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Stabilisatie: Stabilised with polyvinylpyrrolidon (PVP)
M-Clarity™ quality level = MQ300
1.00567.1000 1.00567.2500
1.00567.1000EA 81 EUR
1.00567.1000 1.00567.2500
Lugol's iodine solution stabilised for Gram staining, Sigma-Aldrich®
Lugol's iodine solution
Lugol’s solution stabilized with PVP is used in bacteriology, for example, for the staining of Gram bacteria
It contains a complex of Iodine-Polyvinylpyrrolidon (PVP) and no free iodine, which stabilizes the solution and reduces the evaporation of iodine. During Gram-staining, the iodine from the Lugol’s solution and crystal violet form a stable complex that will not be released by decolorisation. Thus, Gram-positive bacteria will appear dark blue when analyzed microscopically, wheras Gram-negative bacteria can be counterstained e.g. orange with safranin. The Lugol’s solution is registered and quality controlled as IVD / CE product and can be used in diagnostics. 1 L of the Lugol's solution stabilized with PVP is capable of staining up to 500 slides.
IVDLogo
Order Now
Specification Test Results
Suitability for microscopy passes test
gram-positve microorganisms blue-violet
gram-negative microorganisms pink to red