Ghost Low ESD Safety Shoes, Lace-Up
Leverancier: ELTEN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
72071/45 72071/46 72071/47 72071/37 72071/38 72071/39 72071/40 72071/41 72071/42 72071/43 72071/44
ELTN72071/45EA 83.9 EUR
ELTN72071/45 ELTN72071/46 ELTN72071/47 ELTN72071/37 ELTN72071/38 ELTN72071/39 ELTN72071/40 ELTN72071/41 ELTN72071/42 ELTN72071/43 ELTN72071/44
Ghost Low ESD Safety Shoes, Lace-Up
Schoenen en laarzen
Leather-free, casual ESD safety shoe made from hydrophobised microfibre, a padded tongue plus a low profile, monodensity PU sole (L10).
  • Fabric lining for natural ventilation
  • Replaceable insole; orthopaedic insoles possible
  • Full-length inlay sole ESD PRO Lady grey
  • Metal-free, puncture resistant midsole
  • Protective steel toecap (200 J)
Ideal for use in areas where exposure to moisture is expected and where there is a risk of penetration from pointed and sharp objects.
Colour: White
Certificaten: EN 61340 (ESD); DGUV 112-191; EN ISO 20345, S3, SRC