Chemical resistant gloves, uvex rubiflex S NB27B
Leverancier: UVEX
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
6027108 6027109 6027110 6027111
UVEX6027108EA 116 EUR
UVEX6027108 UVEX6027109 UVEX6027110 UVEX6027111
Chemical resistant gloves, uvex rubiflex S NB27B
Handschoenen
These lightweight stockinette gloves feature a special full NBR coating and offer a high level of protection against a diverse range of chemical and mechanical risks. With outstanding tactile feel, touch monitors can also be operated while wearing the glove.
  • Outstanding wearer comfort and ergonomic fit
  • Good resistance to grease, mineral oils and many chemicals
  • Extremely high flexibility
  • Good grip in damp and wet areas
Areas of application: Automotive and chemical industry, laboratories, painting work and food processing.
Certificaten: EN 388:2016 (2111X), EN ISO 374-1:2016 Type A (JKNOPT)
Food contact approved
OEKO-TEX® Standard 100