Industrial gloves, uvex unilite 7700
Leverancier: UVEX
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
6058507 6058508 6058509 6058510 6058511
UVEX6058507EA 49.3 EUR
UVEX6058507 UVEX6058508 UVEX6058509 UVEX6058510 UVEX6058511
Industrial gloves, uvex unilite 7700
Handschoenen
These polyamide/elastane gloves with NBR/PU foam coating on palm and fingertips are flexible and robust protective gloves specially designed for mechanical precision and control works.
  • Good mechanical abrasion resistance
  • Good grip in dry and slightly damp to slightly oily areas
  • Very good fit, good tactile feel and high flexibility
Typical applications include: Fine assembly work, precision work, inspection and sorting.
Certificaten: EN 388:2016 (4131X).